Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,4S)-N-BOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
151907-79-8
C11H17NO4
227.26
MFCD00211287
WOUNTSATDZJBLP-SFYZADRCSA-N
1s,4r-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid,--1s,4r-n-boc-4-aminocyclopent-2-enecarboxylic acid,1s,4r-4-tert-butoxycarbonylamino cyclopent-2-enecarboxylic acid,1s,4r---4-boc-amino-2-cyclopentene-1-carboxylic acid,1r,4s-n-boc-1-aminocyclopent-2-ene-4-carboxylic acid,1s,4r-4-tert-butoxycarbonyl amino cyclopent-2-ene-1-carboxylic acid,1s,4r-4-tert-butoxy carbonyl amino cyclopent-2-ene-1-carboxylic acid,pubchem18542,1s,4r-n-boc-4-aminocyclopent-2-enecarboxylic acid
7019283
(1S,4R)-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopent-2-ene-1-carboxylic acid
CC(C)(C)OC(=O)NC1CC(C=C1)C(=O)O
151907-79-8
C11H17NO4
227.26
MFCD00211287
WOUNTSATDZJBLP-SFYZADRCSA-N
1s,4r-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid,--1s,4r-n-boc-4-aminocyclopent-2-enecarboxylic acid,1s,4r-4-tert-butoxycarbonylamino cyclopent-2-enecarboxylic acid,1s,4r---4-boc-amino-2-cyclopentene-1-carboxylic acid,1r,4s-n-boc-1-aminocyclopent-2-ene-4-carboxylic acid,1s,4r-4-tert-butoxycarbonyl amino cyclopent-2-ene-1-carboxylic acid,1s,4r-4-tert-butoxy carbonyl amino cyclopent-2-ene-1-carboxylic acid,pubchem18542,1s,4r-n-boc-4-aminocyclopent-2-enecarboxylic acid
7019283
(1S,4R)-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopent-2-ene-1-carboxylic acid
CC(C)(C)OC(=O)NC1CC(C=C1)C(=O)O
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1g
Enantiomeric Excess 97.5% min. (GC)
Color Beige to Brown
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 227.26
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material (1R, 4S)-N-BOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 95%, (98% e.e.)
Show More Show Less