Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(1R)-(+)-Camphor, 98%
Click to view available options
Quantity:
25 g
100 g
500 g
Description
(1R)-(+)-Camphor is used as a chiral intermediate and auxiliary. It is also used as a skin antipruritic and as an anti-infective agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(1R)-(+)-Camphor is used as a chiral intermediate and auxiliary. It is also used as a skin antipruritic and as an anti-infective agent.
Solubility
Soluble in water (0.1 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
(1R)-(+)-Camphor is used as a chiral intermediate and auxiliary. It is also used as a skin antipruritic and as an anti-infective agent.
Solubility
Soluble in water (0.1 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 175°C to 180°C |
| Density | 0.99 |
| Boiling Point | 204°C |
| Flash Point | 65°C (149°F) |
| Odor | Camphor-like |
| Quantity | 100 g |
| UN Number | UN2717 |
| Beilstein | 2042745 |
| Merck Index | 14,1732 |
| Solubility Information | Soluble in water (0.1g/L at 20°C). |
| Show More |
Safety and Handling
| Hazard Category | H228-H302-H315-H319-H335-H500 |
| Hazard Statement | GHS H Statement H228-H315-H319 Flammable solid. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: CAMPHOR |
| EINECSNumber | 207-355-2 |
| RTECSNumber | EX1260000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction