Alfa Aesar™ (1R)-(-)-Camphorquinone, 98%

Manufacturer: Alfa Aesar™ B2409203

Quantity 1g

 View more versions of this product

Catalog No. AAB2409203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10334-26-6
    Molecular Formula C10H14O2
    Formula Weight 166.22
    MDL Number MFCD00082863
    Beilstein 2327696
    Synonym (1R)-(-)-2,3-Bornanedione
    Chemical Name or Material (1R)-(-)-Camphorquinone

    Specifications

    Formula Weight 166.22
    Molecular Formula C10H14O2
    MDL Number MFCD00082863
    CAS 10334-26-6
    Beilstein 2327696
    Synonym (1R)-(-)-2,3-Bornanedione
    Melting Point 197° to 202°C
    Quantity 1g
    Optical Rotation −102° (c=2 in Ethanol)
    Chemical Name or Material (1R)-(-)-Camphorquinone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit