Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (1R)-(-)-Camphorquinone, 98%

Click to view available options
Quantity:
1g
5g
10334-26-6
C10H14O2
166.22
MFCD00082863
VNQXSTWCDUXYEZ-TYICEKJOSA-N
1r,4s-1,7,7-trimethylnorbornane-2,3-dione,1r,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione,1s,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione
16211789
(4R)-4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
CC1(C2CCC1(C(=O)C2=O)C)C
10334-26-6
C10H14O2
166.22
MFCD00082863
VNQXSTWCDUXYEZ-TYICEKJOSA-N
1r,4s-1,7,7-trimethylnorbornane-2,3-dione,1r,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione,1s,4r-4,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione
16211789
(4R)-4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
CC1(C2CCC1(C(=O)C2=O)C)C
Quantity 5g
Melting Point 197°C to 202°C
Optical Rotation −102° (c=2 in Ethanol)
Beilstein 2327696
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material (1R)-(-)-Camphorquinone