Learn More
(1R)-endo-(+)-Fenchyl alcohol, 96%, Thermo Scientific Chemicals
Description
(1R)-endo-(+)-Fenchyl alcohol can be extensively used in daily chemicals such as fair antiperspirant, textile softener and liquid detergent. It can also be esterified by different kinds of organic acids to expand its usage in flavors and fragrances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(1R)-endo-(+)-Fenchyl alcohol can be extensively used in daily chemicals such as fair antiperspirant, textile softener and liquid detergent. It can also be esterified by different kinds of organic acids to expand its usage in flavors and fragrances.
Solubility
Insoluble in water.
Notes
Store in cool dry place. Incompatible with oxidizing agents. Ensure good ventilation.
Specifications
Melting Point | 37°C to 44°C |
Molecular Formula | C4H4N2O5 |
Beilstein | 1850917 |
Synonym | barbituric acid, 5,5-dihydroxy, 2,4,6 1h,3h,5h-pyrimidinetrione, 5,5-dihydroxy, unii-o2aap9f8b6, 5,5-dihydroxybarbituric acid, ccris 5957, mesoxalylurea monohydrate, 2,4,5,6 1h,3h-pyrimidinetetrone hydrate, 5,5-dihydroxyperhydropyrimidinetrione, 5,5-dihydroxy-2,4,6 1h,3h,5h-pyrimidinetrione, o2aap9f8b6 |
Solubility Information | Insoluble in water. |
InChI Key | ZIIHZVKHFWOENY-UHFFFAOYSA-N |
SMILES | C1(=O)C(C(=O)NC(=O)N1)(O)O |
IUPAC Name | 5,5-dihydroxy-1,3-diazinane-2,4,6-trione |
Molecular Weight (g/mol) | 160.085 |
PubChem CID | 312231 |
Show More |
Safety and Handling
Hazard Category | H315-H319 |
Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
EINECSNumber | 216-639-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.