Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R)-endo-(+)-Fenchyl alcohol, 96%, Thermo Scientific Chemicals

Catalog Number p-7043870
Click to view available options
Quantity:
100 g
500 g
2217-02-9
MFCD00003760
2217-02-9
MFCD00003760

(1R)-endo-(+)-Fenchyl alcohol can be extensively used in daily chemicals such as fair antiperspirant, textile softener and liquid detergent. It can also be esterified by different kinds of organic acids to expand its usage in flavors and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R)-endo-(+)-Fenchyl alcohol can be extensively used in daily chemicals such as fair antiperspirant, textile softener and liquid detergent. It can also be esterified by different kinds of organic acids to expand its usage in flavors and fragrances.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Incompatible with oxidizing agents. Ensure good ventilation.
TRUSTED_SUSTAINABILITY
Melting Point 37°C to 44°C
Molecular Formula C4H4N2O5
Beilstein 1850917
Synonym barbituric acid, 5,5-dihydroxy, 2,4,6 1h,3h,5h-pyrimidinetrione, 5,5-dihydroxy, unii-o2aap9f8b6, 5,5-dihydroxybarbituric acid, ccris 5957, mesoxalylurea monohydrate, 2,4,5,6 1h,3h-pyrimidinetetrone hydrate, 5,5-dihydroxyperhydropyrimidinetrione, 5,5-dihydroxy-2,4,6 1h,3h,5h-pyrimidinetrione, o2aap9f8b6
Solubility Information Insoluble in water.
InChI Key ZIIHZVKHFWOENY-UHFFFAOYSA-N
SMILES C1(=O)C(C(=O)NC(=O)N1)(O)O
IUPAC Name 5,5-dihydroxy-1,3-diazinane-2,4,6-trione
Molecular Weight (g/mol) 160.085
PubChem CID 312231
Formula Weight 154.25
Percent Purity 96%
Quantity 100 g
Chemical Name or Material (1R)-endo-(+)-Fenchyl alcohol
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 216-639-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit