Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R)-(+)-α-Pinene 98% (≥80% e.e.), ACROS Organics™

Click to view available options
Quantity:
100g
250g
500g
Packaging:
Glass bottle
7785-70-8
C10H16
136.24
MFCD00001346
GRWFGVWFFZKLTI-RKDXNWHRSA-N
+-alpha-pinene,1r,5r-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene,1r-+-alpha-pinene,1r,5r-alpha-pinene,unii-h6cm4twh1w,+-pinene,h6cm4twh1w,1r,5r-2-pinene,bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1r,5r,+-pin-2 3-ene
82227
CHEBI:28261
(1R,5R)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene
CC1=CCC2CC1C2(C)C
7785-70-8
C10H16
136.24
MFCD00001346
GRWFGVWFFZKLTI-RKDXNWHRSA-N
+-alpha-pinene,1r,5r-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene,1r-+-alpha-pinene,1r,5r-alpha-pinene,unii-h6cm4twh1w,+-pinene,h6cm4twh1w,1r,5r-2-pinene,bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1r,5r,+-pin-2 3-ene
82227
CHEBI:28261
(1R,5R)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene
CC1=CCC2CC1C2(C)C
Infrared Spectrum Authentic
Enantiomeric Excess 75% min.
Specific Gravity 0.86
Packaging Glass bottle
Refractive Index 1.4650 to 1.4670
Quantity 500g
Color Colorless
Boiling Point 155.0°C
Melting Point -62.0°C
Flash Point 33°C
Density 0.8600g/mL
Specific Rotation Condition +43.50° (20°C neat)
Specific Rotation + 43.50
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 05, 146
Fieser 15,201
Merck Index 14, 7445
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,chloroform,diethyl ether
Formula Weight 136.24
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material (1R)-(+)-α-Pinene, 98% (80+% e.e.)
Show More Show Less

GHS H Statement
Causes serious eye irritation. Harmful if inhaled. Harmful if swallowed. May be fatal if swallowed and enters airways. Causes skin irritation. Toxic to aquatic life with long lasting effects. May cause an allergic skin reaction. Flammable liquid and vapor.

GHS P Statement
Wear eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger

EINECSNumber : 232-087-8

TSCA : TSCA