Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (1S)-10-Camphorsulfonamide, 97%

Manufacturer:  Alfa Aesar™ 4436114

Quantity 25g

 View more versions of this product

Catalog No. AA4436114


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

60933-63-3
231.32
(1S)-10-Camphorsulfonamide
C10H17NO3S
MFCD00075611

Specifications

60933-63-3
C10H17NO3S
MFCD00075611
25g
97%
Powder
231.32
133° to 135°C (dec.)
(1S)-10-Camphorsulfonamide
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit