Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,2R)-(+)-2-Amino-1-cyclopentanecarboxylic acid hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
50mg
Packaging:
Glass Bottle
128052-92-6
C6H12ClNO2
165.617
MFCD02181205
LVBDVNLIEHCCTP-UYXJWNHNSA-N
1s,2r-2-aminocyclopentanecarboxylic acid hydrochloride,1s,2r-+-2-amino-1-cyclopentanecarboxylic acid hydrochloride,cyclopentanecarboxylic acid, 2-amino-, hydrochloride, 1s,2r,1s,2r-2-aminocyclopentane-1-carboxylic acid hydrochloride,pubchem23256,1s,2s---2-amino-1-cyclopentanecarboxylic acid hydrochloride,cyclopentanecarboxylicacid, 2-amino-, hydrochloride, 1r-trans-9ci,1s,2r-2-amino cyclopetanecarboxylic acid hydrochloride,1s,2r-2-amino-cyclopentanecarboxylic acid hydrochloride,+-1s,2r-2-amino-1-cyclopentane-carboxylic acid hydrochloride
13831733
(1S,2R)-2-aminocyclopentane-1-carboxylic acid;hydrochloride
C1CC(C(C1)N)C(=O)O.Cl
128052-92-6
C6H12ClNO2
165.617
MFCD02181205
LVBDVNLIEHCCTP-UYXJWNHNSA-N
1s,2r-2-aminocyclopentanecarboxylic acid hydrochloride,1s,2r-+-2-amino-1-cyclopentanecarboxylic acid hydrochloride,cyclopentanecarboxylic acid, 2-amino-, hydrochloride, 1s,2r,1s,2r-2-aminocyclopentane-1-carboxylic acid hydrochloride,pubchem23256,1s,2s---2-amino-1-cyclopentanecarboxylic acid hydrochloride,cyclopentanecarboxylicacid, 2-amino-, hydrochloride, 1r-trans-9ci,1s,2r-2-amino cyclopetanecarboxylic acid hydrochloride,1s,2r-2-amino-cyclopentanecarboxylic acid hydrochloride,+-1s,2r-2-amino-1-cyclopentane-carboxylic acid hydrochloride
13831733
(1S,2R)-2-aminocyclopentane-1-carboxylic acid;hydrochloride
C1CC(C(C1)N)C(=O)O.Cl
Packaging Glass Bottle
Color White
Quantity 50mg
Specific Rotation +4.5° to +5.5° (20°C, 589nm) (C=1, Water)
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 165.62
Physical Form Powder
Percent Purity 98%
Chemical Name or Material (1S, 2R)-(+)-2-Amino-1-cyclopentanecarboxylic acid hydrochloride
Show More Show Less