Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,2R,4R)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
250mg
500mg
321744-23-4
C12H21NO5
259.302
MFCD02259719
SSATZBUCOTXXGA-HRDYMLBCSA-N
1r,3s,4r-methyl 3-tert-butoxycarbonyl amino-4-hydroxycyclopentanecarboxylate,methyl 1r,3s,4r-3-tert-butoxycarbonyl amino-4-hydroxycyclopentane-1-carboxylate,1s,2r,4r-n-boc-1-amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester,3alpha-tert-butyloxycarbonyl amino-4alpha-hydroxycyclopentane-1alpha-carboxylic acid methyl ester,cyclopentanecarboxylic acid, 3-1,1-dimethylethoxy carbonyl amino-4-hydroxy-, methyl ester, 1r,3s,4r-9ci,cyclopentanecarboxylicacid, 3-1,1-dimethylethoxy carbonyl amino-4-hydroxy-, methyl ester, 1r,3s,4r,methyl 1r,3r,4s-3-hydroxy-4-2-methylpropan-2-yl oxycarbonylamino cyclopentane-1-carboxylate
7022355
methyl (1R,3R,4S)-3-hydroxy-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylate
CC(C)(C)OC(=O)NC1CC(CC1O)C(=O)OC
321744-23-4
C12H21NO5
259.302
MFCD02259719
SSATZBUCOTXXGA-HRDYMLBCSA-N
1r,3s,4r-methyl 3-tert-butoxycarbonyl amino-4-hydroxycyclopentanecarboxylate,methyl 1r,3s,4r-3-tert-butoxycarbonyl amino-4-hydroxycyclopentane-1-carboxylate,1s,2r,4r-n-boc-1-amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester,3alpha-tert-butyloxycarbonyl amino-4alpha-hydroxycyclopentane-1alpha-carboxylic acid methyl ester,cyclopentanecarboxylic acid, 3-1,1-dimethylethoxy carbonyl amino-4-hydroxy-, methyl ester, 1r,3s,4r-9ci,cyclopentanecarboxylicacid, 3-1,1-dimethylethoxy carbonyl amino-4-hydroxy-, methyl ester, 1r,3s,4r,methyl 1r,3r,4s-3-hydroxy-4-2-methylpropan-2-yl oxycarbonylamino cyclopentane-1-carboxylate
7022355
methyl (1R,3R,4S)-3-hydroxy-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylate
CC(C)(C)OC(=O)NC1CC(CC1O)C(=O)OC
Infrared Spectrum Authentic
Color White to Yellow
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (GC)
Quantity 250mg
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 259.3
Physical Form Moist Crystals
Percent Purity ≥94%
Chemical Name or Material (1S, 2R, 4R)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester, 95%, (98% e.e.)
Show More Show Less