Alfa Aesar™ (1S,2R)-(-)-cis-1-Amino-2-indanol, 97%

Manufacturer: Alfa Aesar™ H3206603

Quantity 1g

 View more versions of this product

Catalog No. AAH3206603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 126456-43-7
    Molecular Formula C9H11NO
    Formula Weight 149.19
    MDL Number MFCD00216655
    UN Number UN3259
    Synonym (1S,2R)-(-)-cis-1-Amino-2-hydroxyindane
    Chemical Name or Material (1S,2R)-(-)-cis-1-Amino-2-indanol

    Specifications

    Formula Weight 149.19
    UN Number UN3259
    Synonym (1S,2R)-(-)-cis-1-Amino-2-hydroxyindane
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 1g
    Optical Rotation −61° (c=0.5 in Chloroform)
    Molecular Formula C9H11NO
    MDL Number MFCD00216655
    CAS 126456-43-7
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Melting Point 117° to 118°C
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
    RTECS Number NK7525500
    Chemical Name or Material (1S,2R)-(-)-cis-1-Amino-2-indanol
    Assay 97%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit