Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
2964-48-9
C9H12N2O4
212.2
MFCD00007359
OCYJXSUPZMNXEN-IUCAKERBSA-N
1s,2s-+-2-amino-1-4-nitrophenyl-1,3-propanediol,1s,2s-2-amino-1-4-nitrophenyl propane-1,3-diol,1s,2s-2-amino-1-4-nitrophenyl-1,3-propanediol,2-amino-1-4-nitro-phenyl-propane-1,3-diol,1,3-propanediol, 2-amino-1-4-nitrophenyl-, 1s,2s,threomine,1s,2s-2-azanyl-1-4-nitrophenyl propane-1,3-diol,1s,2s-+-2-amino-1-4-nitrophenyl-1,3-propaned,1,3-propanediol,2-amino-1-4-nitrophenyl-, 1s,2s,l-threo-+-2-amino-1-4-nitrophenyl-1,3-propanediol
224171
C1=CC(=CC=C1C(C(CO)N)O)[N+](=O)[O-]
2964-48-9
C9H12N2O4
212.2
MFCD00007359
OCYJXSUPZMNXEN-IUCAKERBSA-N
1s,2s-+-2-amino-1-4-nitrophenyl-1,3-propanediol,1s,2s-2-amino-1-4-nitrophenyl propane-1,3-diol,1s,2s-2-amino-1-4-nitrophenyl-1,3-propanediol,2-amino-1-4-nitro-phenyl-propane-1,3-diol,1,3-propanediol, 2-amino-1-4-nitrophenyl-, 1s,2s,threomine,1s,2s-2-azanyl-1-4-nitrophenyl propane-1,3-diol,1s,2s-+-2-amino-1-4-nitrophenyl-1,3-propaned,1,3-propanediol,2-amino-1-4-nitrophenyl-, 1s,2s,l-threo-+-2-amino-1-4-nitrophenyl-1,3-propanediol
224171
C1=CC(=CC=C1C(C(CO)N)O)[N+](=O)[O-]
Melting Point 160°C to 166°C
Infrared Spectrum Authentic
Specific Rotation + 31
Color Yellow to Beige
Quantity 5g
Packaging Glass bottle
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Formula Weight 212.2
Physical Form Crystalline Powder
Percent Purity 96%
Chemical Name or Material (1S, 2S)-2-Amino-1-(4-nitrophenyl)propane-1, 3-diol
Show More Show Less

EINECSNumber : 221-001-4