Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,2S)-2-Amino-1-phenylpropyldiphenylphosphine, 97%, ACROS Organics™

Click to view available options
Quantity:
100mg
250mg
Packaging:
Glass bottle
341968-71-6
C21H22NP
319.38
JWZAIGGNEGTDMG-LAUBAEHRSA-N
1s,2s-1-diphenylphosphino-1-phenylpropan-2-amine,1s,2s-2-amino-1-phenylpropyl diphenylphosphine,1s,2s-2-amino-1-phenylpropyldiphenylphosphine,1s,2s-1-diphenylphosphanyl-1-phenylpropan-2-amine,1s,2s-2-diphenylphosphino-1-methyl-2-phenylethylamine,1s,2s-2-amino-1-phenylpropyl diphenylphosphane,1s,2s-1-diphenylphosphino-1-phenyl-2-propanamine,1s,2s-2-amino-1-phenylpropyldiphenylphosphine,min,s,s-+-2-amino-1-phenylpropyldiphenylphosphine,1s,2s-2-amino-1-phenylpropyl diphenylphosphine, kanata purity
46176707
(1S,2S)-1-diphenylphosphanyl-1-phenylpropan-2-amine
CC(C(C1=CC=CC=C1)P(C2=CC=CC=C2)C3=CC=CC=C3)N
341968-71-6
C21H22NP
319.38
JWZAIGGNEGTDMG-LAUBAEHRSA-N
1s,2s-1-diphenylphosphino-1-phenylpropan-2-amine,1s,2s-2-amino-1-phenylpropyl diphenylphosphine,1s,2s-2-amino-1-phenylpropyldiphenylphosphine,1s,2s-1-diphenylphosphanyl-1-phenylpropan-2-amine,1s,2s-2-diphenylphosphino-1-methyl-2-phenylethylamine,1s,2s-2-amino-1-phenylpropyl diphenylphosphane,1s,2s-1-diphenylphosphino-1-phenyl-2-propanamine,1s,2s-2-amino-1-phenylpropyldiphenylphosphine,min,s,s-+-2-amino-1-phenylpropyldiphenylphosphine,1s,2s-2-amino-1-phenylpropyl diphenylphosphine, kanata purity
46176707
(1S,2S)-1-diphenylphosphanyl-1-phenylpropan-2-amine
CC(C(C1=CC=CC=C1)P(C2=CC=CC=C2)C3=CC=CC=C3)N
Melting Point 100.0°C to 105.0°C
Quantity 250mg
Packaging Glass bottle
NMR 96.0% min.
Color White to Yellow
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 319.38
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material (1S, 2S)-2-Amino-1-phenylpropyldiphenylphosphine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning