Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,2S)-2-Aminocyclohexanecarboxylic Acid 98.0+%, TCI America™

Click to view available options
Quantity:
100mg
24716-93-6
C7H13NO2
143.186
USQHEVWOPJDAAX-WDSKDSINSA-N
1s,2s-2-aminocyclohexanecarboxylic acid,1s,2s-2-aminocyclohexane-1-carboxylic acid,cyclohexanecarboxylic acid, 2-amino-, 1s,2s,1s,2s-2-aminocyclohexanecarboxylicacid,1s,2s-2-amino-cyclohexanecarboxylic acid,2beta-amino-1alpha-cyclohexanecarboxylic acid,cyclohexanecarboxylic acid,2-amino-, 1s,2s,24716-92-5 1s,2s-2-aminocyclohexane-1-carboxylic acid
7128323
(1S,2S)-2-aminocyclohexane-1-carboxylic acid
C1CCC(C(C1)C(=O)O)N
24716-93-6
C7H13NO2
143.186
USQHEVWOPJDAAX-WDSKDSINSA-N
1s,2s-2-aminocyclohexanecarboxylic acid,1s,2s-2-aminocyclohexane-1-carboxylic acid,cyclohexanecarboxylic acid, 2-amino-, 1s,2s,1s,2s-2-aminocyclohexanecarboxylicacid,1s,2s-2-amino-cyclohexanecarboxylic acid,2beta-amino-1alpha-cyclohexanecarboxylic acid,cyclohexanecarboxylic acid,2-amino-, 1s,2s,24716-92-5 1s,2s-2-aminocyclohexane-1-carboxylic acid
7128323
(1S,2S)-2-aminocyclohexane-1-carboxylic acid
C1CCC(C(C1)C(=O)O)N
Color White
Quantity 100mg
Formula Weight 143.19
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material (1S,2S)-2-Aminocyclohexanecarboxylic Acid