Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,2S,4R)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
500mg
262280-14-8
C12H21NO5
259.302
MFCD02259721
SSATZBUCOTXXGA-VGMNWLOBSA-N
1r,3s,4s-methyl 3-tert-butoxycarbonyl amino-4-hydroxycyclopentanecarboxylate,1s,2s,4r-n-boc-1-amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester,methyl 1r,3s,4s-3-tert-butoxycarbonyl amino-4-hydroxycyclopentane-1-carboxylate,methyl 1r,3s,4s-3-t-butoxy carbonyl amino-4-hydroxycyclopentane-1-carboxylate,3alpha-tert-butyloxycarbonyl amino-4beta-hydroxycyclopentane-1alpha-carboxylic acid methyl ester,cyclopentanecarboxylic acid, 3-1,1-dimethylethoxy carbonyl amino-4-hydroxy-, methyl ester, 1r,3s,4s-9ci,cyclopentanecarboxylicacid, 3-1,1-dimethylethoxy carbonyl amino-4-hydroxy-, methyl ester, 1r,3s,4s,methyl 1r,3s,4s-3-hydroxy-4-2-methylpropan-2-yl oxycarbonylamino cyclopentane-1-carboxylate
7022354
methyl (1R,3S,4S)-3-hydroxy-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylate
CC(C)(C)OC(=O)NC1CC(CC1O)C(=O)OC
262280-14-8
C12H21NO5
259.302
MFCD02259721
SSATZBUCOTXXGA-VGMNWLOBSA-N
1r,3s,4s-methyl 3-tert-butoxycarbonyl amino-4-hydroxycyclopentanecarboxylate,1s,2s,4r-n-boc-1-amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester,methyl 1r,3s,4s-3-tert-butoxycarbonyl amino-4-hydroxycyclopentane-1-carboxylate,methyl 1r,3s,4s-3-t-butoxy carbonyl amino-4-hydroxycyclopentane-1-carboxylate,3alpha-tert-butyloxycarbonyl amino-4beta-hydroxycyclopentane-1alpha-carboxylic acid methyl ester,cyclopentanecarboxylic acid, 3-1,1-dimethylethoxy carbonyl amino-4-hydroxy-, methyl ester, 1r,3s,4s-9ci,cyclopentanecarboxylicacid, 3-1,1-dimethylethoxy carbonyl amino-4-hydroxy-, methyl ester, 1r,3s,4s,methyl 1r,3s,4s-3-hydroxy-4-2-methylpropan-2-yl oxycarbonylamino cyclopentane-1-carboxylate
7022354
methyl (1R,3S,4S)-3-hydroxy-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylate
CC(C)(C)OC(=O)NC1CC(CC1O)C(=O)OC
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Quantity 500mg
Color White
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 259.3
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material (1S, 2S, 4R)-N-BOC-1-Amino-2-hydroxycyclopentane-4-carboxylic acid methyl ester, 95%, (98% e.e.)
Show More Show Less