Alfa Aesar™ (1S,2S)-Bis(4-methoxyphenyl)-1,2-ethanediamine, 98%

Manufacturer: Alfa Aesar™ H2580114

Quantity 25g

 View more versions of this product

Catalog No. AAH2580114

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 58520-04-0
    Molecular Formula C16H20N2O2
    Formula Weight 272.34
    MDL Number MFCD08460191
    Synonym (1S,2S)-Bis(2-methoxyphenyl)-1,2-diaminoethane; (S,S)-DAEN
    Chemical Name or Material (1S,2S)-Bis(4-methoxyphenyl)-1,2-ethanediamine

    Specifications

    Formula Weight 272.34
    MDL Number MFCD08460191
    Melting Point 89° to 91°C
    Molecular Formula C16H20N2O2
    CAS 58520-04-0
    Synonym (1S,2S)-Bis(2-methoxyphenyl)-1,2-diaminoethane; (S,S)-DAEN
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 25g
    Optical Rotation −77° (c=2 in Chloroform)
    Chemical Name or Material (1S,2S)-Bis(4-methoxyphenyl)-1,2-ethanediamine
    Assay 98%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit