Alfa Aesar™ (1S,3R)-(+)-3-(Boc-amino)cyclopentanecarboxylic acid, 95%

Manufacturer: Alfa Aesar™ H52194MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52194MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 261165-05-3
    Molecular Formula C11H19NO4
    Formula Weight 229.28
    MDL Number MFCD01320857
    Chemical Name or Material (1S,3R)-(+)-3-(Boc-amino)cyclopentanecarboxylic acid

    Specifications

    Formula Weight 229.28
    Molecular Formula C11H19NO4
    MDL Number MFCD01320857
    CAS 261165-05-3
    Melting Point 110° to 114°C
    Quantity 250mg
    Optical Rotation +16.0° (c=1 in Methanol)
    Chemical Name or Material (1S,3R)-(+)-3-(Boc-amino)cyclopentanecarboxylic acid
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit