Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (1S,3R)-(-)-Camphoric acid, 98%

Manufacturer:  Alfa Aesar™ H5404203

Quantity 1g

 View more versions of this product

Catalog No. AAH5404203


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

560-09-8
200.23
1874674
(1S,3R)-(-)-Camphoric acid
C10H16O4
MFCD00064937
14,1733

Specifications

560-09-8
200.23
1874674
188° to 190°C
−48° (c=10 in Ethanol)
98%
C10H16O4
MFCD00064937
14,1733
1g
(1S,3R)-(-)-Camphoric acid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit