Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,3S)-N-BOC-1-Aminocyclopentane-3-carboxylic Acid Methyl Ester 95%, (98% e.e.), ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
329910-39-6
C12H21NO4
243.303
ZZGMDDXYOHHJMT-IUCAKERBSA-N
methyl 1s,3s-3-tert-butoxy carbonyl amino cyclopentane-1-carboxylate,methyl 1s,3s-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylate,1s,3s-n-boc-1-aminocyclopentane-3-carboxylic acid methyl ester,1s,3s-methyl n-boc-1-aminocyclopentane-3-carboxylate,1s,3s-n-boc-1-aminocyclopentane-3-carboxylic aci,1s, 3s-n-boc-1-aminocyclopentane-3-carboxylic acid methyl ester,1s,3s-1-boc-amino cyclopentane-3-carboxylic acid methyl ester,1s,3s-3-tert-butoxycarbonylamino-cyclopentanecarboxylic acid methyl ester,1s,3s-3-tert-butoxycarbonylaminocyclopentanecarboxylic acid methyl ester,1s,3s-methyl 3-tert-butoxycarbonyl amino cyclopentanecarboxylate
2734531
methyl (1S,3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylate
CC(C)(C)OC(=O)NC1CCC(C1)C(=O)OC
329910-39-6
C12H21NO4
243.303
ZZGMDDXYOHHJMT-IUCAKERBSA-N
methyl 1s,3s-3-tert-butoxy carbonyl amino cyclopentane-1-carboxylate,methyl 1s,3s-3-tert-butoxycarbonyl amino cyclopentane-1-carboxylate,1s,3s-n-boc-1-aminocyclopentane-3-carboxylic acid methyl ester,1s,3s-methyl n-boc-1-aminocyclopentane-3-carboxylate,1s,3s-n-boc-1-aminocyclopentane-3-carboxylic aci,1s, 3s-n-boc-1-aminocyclopentane-3-carboxylic acid methyl ester,1s,3s-1-boc-amino cyclopentane-3-carboxylic acid methyl ester,1s,3s-3-tert-butoxycarbonylamino-cyclopentanecarboxylic acid methyl ester,1s,3s-3-tert-butoxycarbonylaminocyclopentanecarboxylic acid methyl ester,1s,3s-methyl 3-tert-butoxycarbonyl amino cyclopentanecarboxylate
2734531
methyl (1S,3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylate
CC(C)(C)OC(=O)NC1CCC(C1)C(=O)OC
Infrared Spectrum Authentic
Specific Rotation +8° to +9° (20°C, 589nm) (c=1, methanol)
Packaging Glass bottle
Color White
Quantity 500mg
Enantiomeric Excess 97.5% min.
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 243.3
Physical Form Needle-Like Crystals
Percent Purity ≥94%
Chemical Name or Material (1S, 3S)-N-BOC-1-Aminocyclopentane-3-carboxylic acid methyl ester, 95%, (98% e.e.)
Show More Show Less