Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,4R)-2-Azabicyclo[2.2.1]heptan-3-one, 95%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
134003-03-5
C6H9NO
111.14
UIVLZOWDXYXITH-UHNVWZDZSA-N
1s,4r-2-azabicyclo 2.2.1 heptan-3-one,2-azabicyclo 2.2.1 heptan-3-one, 1s,4r,1s,4r-2-aza-bicyclo 2.2.1 heptan-3-one,1beta,4beta-2-azabicyclo 2.2.1 heptane-3-one
2734523
(1R,4S)-3-azabicyclo[2.2.1]heptan-2-one
C1CC2CC1C(=O)N2
134003-03-5
C6H9NO
111.14
UIVLZOWDXYXITH-UHNVWZDZSA-N
1s,4r-2-azabicyclo 2.2.1 heptan-3-one,2-azabicyclo 2.2.1 heptan-3-one, 1s,4r,1s,4r-2-aza-bicyclo 2.2.1 heptan-3-one,1beta,4beta-2-azabicyclo 2.2.1 heptane-3-one
2734523
(1R,4S)-3-azabicyclo[2.2.1]heptan-2-one
C1CC2CC1C(=O)N2
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250mg
Color White to Yellow
Enantiomeric Excess 97.5% min.
Melting Point 76.2°C
Assay Percent Range 94% min. (GC)
Formula Weight 111.14
Physical Form Crystalline Powder
Percent Purity 95%,98% ee
Chemical Name or Material (1S,4R)-2-Azabicyclo[2.2.1]heptan-3-one
Show More Show Less