Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,4R)-N-BOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 95%, 98% ee, Acros Organics

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
151907-80-1
C11H17NO4
227.26
MFCD00211288
WOUNTSATDZJBLP-JGVFFNPUSA-N
1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid,1r,4s-4-tert-butoxycarbonylamino cyclopent-2-enecarboxylic acid,1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxylic acid,+-1r,4s-n-boc-4-aminocyclopent-2-enecarboxylic acid,1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-ene-1-carboxylic acid,+-1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxy,cis-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid,1r,4s-4-tert-butoxy carbonyl amino cyclopent-2-ene-1-carboxylic acid,1r,4s-rel-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid
7019281
(1R,4S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopent-2-ene-1-carboxylic acid
CC(C)(C)OC(=O)NC1CC(C=C1)C(=O)O
151907-80-1
C11H17NO4
227.26
MFCD00211288
WOUNTSATDZJBLP-JGVFFNPUSA-N
1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid,1r,4s-4-tert-butoxycarbonylamino cyclopent-2-enecarboxylic acid,1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxylic acid,+-1r,4s-n-boc-4-aminocyclopent-2-enecarboxylic acid,1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-ene-1-carboxylic acid,+-1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxy,cis-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid,1r,4s-4-tert-butoxy carbonyl amino cyclopent-2-ene-1-carboxylic acid,1r,4s-rel-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid
7019281
(1R,4S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopent-2-ene-1-carboxylic acid
CC(C)(C)OC(=O)NC1CC(C=C1)C(=O)O
Melting Point 152°C
Packaging Glass bottle
Quantity 250mg
Infrared Spectrum Authentic
Color White
Enantiomeric Excess 97.5% min. (HPLC)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Solubility Information Solubility in water: soluble
Formula Weight 227.26
Physical Form Crystalline Powder
Percent Purity 95%,98% ee
Chemical Name or Material (1S, 4R)-N-BOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 98% ee
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning