Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane, 97%, Alfa Aesar™

Click to view available options
Quantity:
250mg
1g
5g
113451-59-5
C10H18N2O2
198.266
MFCD01569250
UXAWXZDXVOYLII-YUMQZZPRSA-N
1s,4s-2-boc-2,5-diazabicyclo 2.2.1 heptane,tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylate,1s,4s-tert-butyl 2,5-diazabicyclo 2.2.1 heptane-2-carboxylate,1s,4s-n-tert-butoxycarbonyl-2,5-diazabicyclo 2.2.1 heptane,1s,4s---2-boc-2,5-diazabicyclo 2.2.1 heptane,tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptan-2-carboxylate,1s,4s-2,5-diaza-bicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester,1s,4s-n-boc-2,5-diazabicyclo 2.2.1 heptane,1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester,tert-butyl 1s,4s---2,5-diazabicyclo 2.2.1 heptane-2-carboxylate
11521263
tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
CC(C)(C)OC(=O)N1CC2CC1CN2
113451-59-5
C10H18N2O2
198.266
MFCD01569250
UXAWXZDXVOYLII-YUMQZZPRSA-N
1s,4s-2-boc-2,5-diazabicyclo 2.2.1 heptane,tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylate,1s,4s-tert-butyl 2,5-diazabicyclo 2.2.1 heptane-2-carboxylate,1s,4s-n-tert-butoxycarbonyl-2,5-diazabicyclo 2.2.1 heptane,1s,4s---2-boc-2,5-diazabicyclo 2.2.1 heptane,tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptan-2-carboxylate,1s,4s-2,5-diaza-bicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester,1s,4s-n-boc-2,5-diazabicyclo 2.2.1 heptane,1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester,tert-butyl 1s,4s---2,5-diazabicyclo 2.2.1 heptane-2-carboxylate
11521263
tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
CC(C)(C)OC(=O)N1CC2CC1CN2
Applications
It is used as an active pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from air, carbon dioxide, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.
Sensitivity Air sensitive
Quantity 250mg
Optical Rotation −44° (c=1 in Chloroform)
Melting Point 74°C to 76°C
Beilstein 6460311
Solubility Information Insoluble in water.
Formula Weight 198.26
Percent Purity 97%
Chemical Name or Material (1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen