Alfa Aesar™ (1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane, 98%

Manufacturer: Alfa Aesar™ H5109703

Quantity 1g

 View more versions of this product

Catalog No. AAH5109703

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 113451-59-5
    Molecular Formula C10H18N2O2
    Formula Weight 198.26
    MDL Number MFCD01569250
    Synonym tert-Butyl(1S,4S)-(-)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
    Chemical Name or Material (1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane

    Specifications

    Formula Weight 198.26
    MDL Number MFCD01569250
    Melting Point 74° to 76°C
    Quantity 1g
    Optical Rotation −44° (c=1 in Chloroform)
    Molecular Formula C10H18N2O2
    CAS 113451-59-5
    Synonym tert-Butyl(1S,4S)-(-)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Recommended Storage 2° to 8°C
    Chemical Name or Material (1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane
    Assay 98%

    Description

    Applications
    It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

    Solubility
    Insoluble in water.

    Notes
    Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit