Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane, 98%

Manufacturer:  Alfa Aesar™ H5109706

Quantity 5g

 View more versions of this product

Catalog No. AAH5109706

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Description & Specifications

Chemical Identifiers

113451-59-5
198.26
tert-Butyl(1S,4S)-(-)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
C10H18N2O2
MFCD01569250
(1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane

Specifications

113451-59-5
198.26
tert-Butyl(1S,4S)-(-)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
5g
(1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane
C10H18N2O2
MFCD01569250
74° to 76°C
−44° (c=1 in Chloroform)
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Warning

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit