Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (1S)-(-)-Camphor, 98%

Manufacturer:  Alfa Aesar™ B2346906

Quantity 5g

 View more versions of this product

Catalog No. AAB2346906


  Check Availability
Add to cart

Specifications

Description

Applications
(1S)-(-)-Camphor is used as chiral intermediate and chiral auxiliary precursor. It is used in the synthesis of high-potency sweeteners. It acts as catalytic agent and petrochemical additive.

Solubility
Soluble in water. (0.34 g/L) at 20°C

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Description & Specifications

Chemical Identifiers

464-48-2
152.24
UN2717
14,1732
(1S)-(-)-Camphor
C10H16O
MFCD00064148
4291747
L-2-Bornanone

Specifications

464-48-2
152.24
UN2717
14,1732
204°C
5g
Camphor-like
(1S)-(-)-Camphor
C10H16O
MFCD00064148
4291747
L-2-Bornanone
175° to 180°C
65°C (149°F)
−43° (c=10 in Ethanol)
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H315-H319
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P280g-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: CAMPHOR

RTECSNumber : EX1250000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit