Alfa Aesar™ (1S)-(-)-Camphor, 98%

Manufacturer: Alfa Aesar™ B2346914

Quantity 25g

 View more versions of this product

Catalog No. AAB2346914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 464-48-2
    Molecular Formula C10H16O
    Formula Weight 152.24
    MDL Number MFCD00064148
    UN Number UN2717
    Beilstein 4291747
    Merck Index 14,1732
    Synonym L-2-Bornanone
    Chemical Name or Material (1S)-(-)-Camphor

    Specifications

    Formula Weight 152.24
    Boiling Point 204°C
    Melting Point 175° to 180°C
    Quantity 25g
    Flash Point 65°C (149°F)
    Odor Camphor-like
    Optical Rotation −43° (c=10 in Ethanol)
    TSCA TSCA
    Molecular Formula C10H16O
    MDL Number MFCD00064148
    UN Number UN2717
    Beilstein 4291747
    Merck Index 14,1732
    Synonym L-2-Bornanone
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H228-H315-H319
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P210-P280g-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: CAMPHOR
    RTECS Number EX1250000
    CAS 464-48-2
    Chemical Name or Material (1S)-(-)-Camphor
    Assay 98%

    Description

    Applications
    (1S)-(-)-Camphor is used as chiral intermediate and chiral auxiliary precursor. It is used in the synthesis of high-potency sweeteners. It acts as catalytic agent and petrochemical additive.

    Solubility
    Soluble in water. (0.34 g/L) at 20°C

    Notes
    Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

    GHS H Statement
    H228-H315-H319
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P210-P280g-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit