Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S)-(-)-β-Pinene, 98%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
5L
Packaging:
Glass bottle
Metal drum
18172-67-3
C10H16
136.24
MFCD00001345
WTARULDDTDQWMU-IUCAKERBSA-N
--beta-pinene,--nopinene,1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane,1s---beta-pinene,--b-pinene,--pin-2 10-ene,unii-afn153a7su,--?-pinene,1s-1-beta-pinene,afn153a7su
440967
CHEBI:28359
(1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
CC1(C2CCC(=C)C1C2)C
18172-67-3
C10H16
136.24
MFCD00001345
WTARULDDTDQWMU-IUCAKERBSA-N
--beta-pinene,--nopinene,1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane,1s---beta-pinene,--b-pinene,--pin-2 10-ene,unii-afn153a7su,--?-pinene,1s-1-beta-pinene,afn153a7su
440967
CHEBI:28359
(1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
CC1(C2CCC(=C)C1C2)C
Specific Rotation -19.5
Specific Gravity 0.87
Quantity 5L
Specific Rotation Condition − 19.50 (25.00°C c=neat)
Flash Point 32°C
Infrared Spectrum Authentic
Density 0.8700g/mL
Color Colorless to Yellow
Refractive Index 1.4770 to 1.4810
Boiling Point 165.0°C to 167.0°C
Packaging Metal drum
Melting Point -61.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
UN Number 2319
Beilstein 05,154
Merck Index 14,7446
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,chloroform and ether
Formula Weight 136.24
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material (1S)-(-)-β-Pinene
Show More Show Less

GHS H Statement:
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid release to the environment.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger