Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1S)-(-)-Verbenone, 94%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
1196-01-6
C10H14O
150.22
MFCD00065445
DCSCXTJOXBUFGB-JGVFFNPUSA-N
levoverbenone,--verbenone,l-verbenone,1s---verbenone,--2-pinen-4-one,verbenone,unii-2xp0j7754u,1s,5s-4,6,6-trimethylbicyclo 3.1.1 hept-3-en-2-one,verbenone, l,bicyclo 3.1.1 hept-3-en-2-one, 4,6,6-trimethyl-, 1s,5s
92874
CHEBI:78316
(1S,5S)-2,6,6-trimethylbicyclo[3.1.1]hept-2-en-4-one
CC1=CC(=O)C2CC1C2(C)C
1196-01-6
C10H14O
150.22
MFCD00065445
DCSCXTJOXBUFGB-JGVFFNPUSA-N
levoverbenone,--verbenone,l-verbenone,1s---verbenone,--2-pinen-4-one,verbenone,unii-2xp0j7754u,1s,5s-4,6,6-trimethylbicyclo 3.1.1 hept-3-en-2-one,verbenone, l,bicyclo 3.1.1 hept-3-en-2-one, 4,6,6-trimethyl-, 1s,5s
92874
CHEBI:78316
(1S,5S)-2,6,6-trimethylbicyclo[3.1.1]hept-2-en-4-one
CC1=CC(=O)C2CC1C2(C)C
Boiling Point 227°C to 228°C
Flash Point 85°C
Refractive Index 1.4950 to 1.497
Specific Rotation -220
Specific Gravity 0.97
Packaging Glass bottle
Specific Rotation Condition − 220.00 (20.00°C c=3−4,CHCL3)
Density 0.9700g/mL
Color Yellow
Quantity 10g
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Beilstein 07, 161
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with most common organic solvents
Formula Weight 150.22
Physical Form Liquid
Percent Purity 94%
Chemical Name or Material (1S)-(-)-Verbenone, 94%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:

GHS Signal Word: Warning

EINECSNumber : 214-807-2

TSCA : TSCA