Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-(1,1-Difluoroethyl)pyrazine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Boiling Point | 150.0°C to 152.0°C |
Flash Point | 53°C |
Assay Percent Range | 96% min. (GC) |
Quantity | 1 g |
Formula Weight | 144.12 |
Percent Purity | 97% |
Chemical Name or Material | 2-(1, 1-Difluoroethyl)pyrazine |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Flammable liquid and vapour. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation. |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction