Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,1,3-Benzothiadiazole-4-sulfonyl chloride, 97%, Thermo Scientific™

Click to view available options
Quantity:
1 g
5 g
73713-79-8
C6H3ClN2O2S2
234.67
MFCD00068049
CXAICGCTHOWKPP-UHFFFAOYSA-N
benzo c 1,2,5 thiadiazole-4-sulfonyl chloride, benzo 2,1,3 thiadiazole-4-sulfonyl chloride, 2,1,3-benzothiadiazole-4-sulphonyl chloride, 2,1,3-benzothiadiazol-4-sulfonyl chloride, 4-chlorosulfonyl-2,1,3-benzothiadiazole, 2,1,3-benzothiadiazole, 4-chlorosulfonyl, 2,1,3-benzothiadiazole-4-sulfonylchloride, benzo-2,1,3-thiadiazole-4-sulfonyl chloride, 2??,1,3-benzothiadiazole-4-sulfonyl chloride, benzo-2,1,3-thiadiazole-4-sulphonyl chloride
2735459
2,1,3-benzothiadiazole-4-sulfonyl chloride
ClS(=O)(=O)C1=CC=CC2=NSN=C12
73713-79-8
C6H3ClN2O2S2
234.67
MFCD00068049
CXAICGCTHOWKPP-UHFFFAOYSA-N
benzo c 1,2,5 thiadiazole-4-sulfonyl chloride, benzo 2,1,3 thiadiazole-4-sulfonyl chloride, 2,1,3-benzothiadiazole-4-sulphonyl chloride, 2,1,3-benzothiadiazol-4-sulfonyl chloride, 4-chlorosulfonyl-2,1,3-benzothiadiazole, 2,1,3-benzothiadiazole, 4-chlorosulfonyl, 2,1,3-benzothiadiazole-4-sulfonylchloride, benzo-2,1,3-thiadiazole-4-sulfonyl chloride, 2??,1,3-benzothiadiazole-4-sulfonyl chloride, benzo-2,1,3-thiadiazole-4-sulphonyl chloride
2735459
2,1,3-benzothiadiazole-4-sulfonyl chloride
ClS(=O)(=O)C1=CC=CC2=NSN=C12

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 145°C to 147°C
Quantity 1 g
UN Number UN3261
Formula Weight 234.68
Percent Purity 97%
Chemical Name or Material 2,1,3-Benzothiadiazole-4-sulfonyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : DL0100000

TSCA : No

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit