Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1,3-Dioxolan-2-yl)ethyltriphenylphosphonium bromide, 97%, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
86608-70-0
C23H24BrO2P
443.321
MFCD00075119
ZCJKBPSRKLHANV-UHFFFAOYSA-M
2-1,3-dioxolan-2-yl ethyltriphenylphosphonium bromide,2-1,3-dioxolan-2-yl ethyl triphenylphosphonium bromide,phosphonium, 2-1,3-dioxolan-2-yl ethyl triphenyl-, bromide,2-1,3-dioxolan-2-yl ethyl triphenylphosphanium bromide,acmc-1bl1m,2-1,3-dioxolan-2-yl ethyltrip,phosphonium, 2-1,3-dioxolan-2-yl ethyl triphenyl-, bromide 1:1,2-1,3-dioxolan-2-yl ethyltriphenylphosp,2-1,3-dioxolan-2-yl ethyltriphenylphosphoniumbromide
2733834
2-(1,3-dioxolan-2-yl)ethyl-triphenylphosphanium;bromide
C1COC(O1)CC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
86608-70-0
C23H24BrO2P
443.321
MFCD00075119
ZCJKBPSRKLHANV-UHFFFAOYSA-M
2-1,3-dioxolan-2-yl ethyltriphenylphosphonium bromide,2-1,3-dioxolan-2-yl ethyl triphenylphosphonium bromide,phosphonium, 2-1,3-dioxolan-2-yl ethyl triphenyl-, bromide,2-1,3-dioxolan-2-yl ethyl triphenylphosphanium bromide,acmc-1bl1m,2-1,3-dioxolan-2-yl ethyltrip,phosphonium, 2-1,3-dioxolan-2-yl ethyl triphenyl-, bromide 1:1,2-1,3-dioxolan-2-yl ethyltriphenylphosp,2-1,3-dioxolan-2-yl ethyltriphenylphosphoniumbromide
2733834
2-(1,3-dioxolan-2-yl)ethyl-triphenylphosphanium;bromide
C1COC(O1)CC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
for C-C Bond Formation (Olefination)
Quantity 5g
Packaging Glass bottle
Color Yellow
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 443.32
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-(1, 3-Dioxolan-2-yl)ethyltriphenylphosphonium bromide, 97%
Show More Show Less

GHS H Statement:
Harmful to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

TSCA : TSCA