Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[1-(4-Piperonyl)piperazinyl]benzothiazole, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
155106-73-3
C19H19N3O2S
353.44
BYHKGNWKJMGHGE-UHFFFAOYSA-N
2-ppbt, 2-1-4-piperonyl piperazinyl benzothiazole, chembl24909, 2-4-1,3-benzodioxol-5-ylmethyl-1-piperazinyl benzothiazole, 2-4-benzo d 1,3 dioxol-5-ylmethyl piperazin-1-yl benzo d thiazole, benzothiazole, 2-4-1,3-benzodioxol-5-ylmethyl-1-piperazinyl, 2-4-2h-1,3-benzodioxol-5-ylmethyl piperazin-1-yl-1,3-benzothiazole, tocris-0736, biomol-nt_000145
127902
2-[4-(1,3-benzodioxol-5-ylmethyl)piperazin-1-yl]-1,3-benzothiazole
C1CN(CCN1CC2=CC3=C(C=C2)OCO3)C4=NC5=CC=CC=C5S4
This item is not returnable. View return policy
155106-73-3
C19H19N3O2S
353.44
BYHKGNWKJMGHGE-UHFFFAOYSA-N
2-ppbt, 2-1-4-piperonyl piperazinyl benzothiazole, chembl24909, 2-4-1,3-benzodioxol-5-ylmethyl-1-piperazinyl benzothiazole, 2-4-benzo d 1,3 dioxol-5-ylmethyl piperazin-1-yl benzo d thiazole, benzothiazole, 2-4-1,3-benzodioxol-5-ylmethyl-1-piperazinyl, 2-4-2h-1,3-benzodioxol-5-ylmethyl piperazin-1-yl-1,3-benzothiazole, tocris-0736, biomol-nt_000145
127902
2-[4-(1,3-benzodioxol-5-ylmethyl)piperazin-1-yl]-1,3-benzothiazole
C1CN(CCN1CC2=CC3=C(C=C2)OCO3)C4=NC5=CC=CC=C5S4

5-HT4 agonist; also 5-HT3 antagonist

5-HT4 receptor agonist with moderate activity as a 5-HT3 antagonist. Has no affinity at 5-HT1 or dopamine D2 receptors. Potential gastrokinetic agent.
Quantity 50mg
Formula Weight 353.44
Percent Purity >98%
Chemical Name or Material 2-[1-(4-Piperonyl)piperazinyl]benzothiazole

Recommended Storage : Store at Room Temperature