Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[1-(4-Piperonyl)piperazinyl]benzothiazole, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

155106-73-3
C19H19N3O2S
353.44
BYHKGNWKJMGHGE-UHFFFAOYSA-N
127902

5-HT4 agonist. Also 5-HT3 antagonist

5-HT4 receptor agonist with moderate activity as a 5-HT3 antagonist. Has no affinity at 5-HT1 or dopamine D2 receptors. Potential gastrokinetic agent.
SMILES C1CN(CCN1CC2=CC3=C(C=C2)OCO3)C4=NC5=CC=CC=C5S4
IUPAC Name 2-[4-(1,3-benzodioxol-5-ylmethyl)piperazin-1-yl]-1,3-benzothiazole
Formula Weight 353.44
Quantity 10mg
Chemical Name or Material 2-[1-(4-Piperonyl)piperazinyl]benzothiazole
Grade >98%

Recommended Storage : Store at Room Temperature