Alfa Aesar™ 2-(1-Boc-4-piperidinyloxy)-N-methylacetamide, 96%

Manufacturer: Alfa Aesar™ H3299003

Quantity 1g

 View more versions of this product

Catalog No. AAH3299003

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C13H24N2O4
    Formula Weight 272.35
    MDL Number MFCD17015029
    Synonym 4-(Methylcarbamoylmethoxy)piperidine-1-carboxylic acid tert-butyl ester
    Chemical Name or Material 2-(1-Boc-4-piperidinyloxy)-N-methylacetamide

    Specifications

    Formula Weight 272.35
    Molecular Formula C13H24N2O4
    MDL Number MFCD17015029
    Melting Point 94° to 96°C
    Synonym 4-(Methylcarbamoylmethoxy)piperidine-1-carboxylic acid tert-butyl ester
    Quantity 1g
    Chemical Name or Material 2-(1-Boc-4-piperidinyloxy)-N-methylacetamide
    Assay 96%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit