Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(1-Imidazolyl)acetonitrile, 95%

Click to view available options
Quantity:
250mg
1g
98873-55-3
C5H5N3
107.116
MFCD06421433
ZPGCVVBPGQJSPX-UHFFFAOYSA-N
2-1h-imidazol-1-yl acetonitrile,imidazol-1-yl-acetonitrile,1-cyanomethyl imidazole,1h-imidazole-1-acetonitrile,1h-imidazol-1-ylacetonitrile,2-1-imidazolyl acetonitrile,1-imidazolyl acetonitrile,2-imidazol-1-yl acetonitrile,1h-imidazol-1-yl acetonitrile,1-cyanomethylimidazole
11804695
2-imidazol-1-ylacetonitrile
C1=CN(C=N1)CC#N
98873-55-3
C5H5N3
107.116
MFCD06421433
ZPGCVVBPGQJSPX-UHFFFAOYSA-N
2-1h-imidazol-1-yl acetonitrile,imidazol-1-yl-acetonitrile,1-cyanomethyl imidazole,1h-imidazole-1-acetonitrile,1h-imidazol-1-ylacetonitrile,2-1-imidazolyl acetonitrile,1-imidazolyl acetonitrile,2-imidazol-1-yl acetonitrile,1h-imidazol-1-yl acetonitrile,1-cyanomethylimidazole
11804695
2-imidazol-1-ylacetonitrile
C1=CN(C=N1)CC#N
Quantity 1g
UN Number UN3439
Beilstein 7473110
Formula Weight 107.12
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-(1-Imidazolyl)acetonitrile

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

storageNote1 : -30° to -10°C