Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1-Naphthyl)ethanol, 95%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
773-99-9
C12H12O
172.23
RXWNCMHRJCOWDK-UHFFFAOYSA-N
1-naphthaleneethanol, 2-1-naphthyl ethanol, 2-naphthalen-1-yl ethanol, 1-2-hydroxyethyl naphthalene, naphthalen-1-ethanol, 2-naphthylethanol, 1-naphthyl ethanol, 1-naphthalene ethanol, 2-naphthalen-1-yl ethan-1-ol, 1-napthaleneethanol
13047
2-naphthalen-1-ylethanol
C1=CC=C2C(=C1)C=CC=C2CCO
773-99-9
C12H12O
172.23
RXWNCMHRJCOWDK-UHFFFAOYSA-N
1-naphthaleneethanol, 2-1-naphthyl ethanol, 2-naphthalen-1-yl ethanol, 1-2-hydroxyethyl naphthalene, naphthalen-1-ethanol, 2-naphthylethanol, 1-naphthyl ethanol, 1-naphthalene ethanol, 2-naphthalen-1-yl ethan-1-ol, 1-napthaleneethanol
13047
2-naphthalen-1-ylethanol
C1=CC=C2C(=C1)C=CC=C2CCO
Boiling Point 186°C (23.0 hPa)
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Quantity 10g
Flash Point 96°C
Melting Point 61.0°C to 65.0°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Linear Formula C10H7CH2CH2OH
Beilstein 06,668
Formula Weight 172.23
Physical Form Solid or Crystalline Powder
Percent Purity 95%
Chemical Name or Material 2-(1-Naphthyl)ethanol
Show More Show Less

EINECSNumber : 212-260-4