Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1-Piperazinyl)pyrimidine, 98+%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
20980-22-7
C8H12N4
164.21
MFCD00040742
MRBFGEHILMYPTF-UHFFFAOYSA-N
2-1-piperazinyl pyrimidine,1-2-pyrimidyl piperazine,2-piperazin-1-yl pyrimidine,1-2-pyrimidinyl piperazine,2-piperazinopyrimidine,campiron,pyrimidine, 2-1-piperazinyl,campirone,kampirone,2-piperazin-1-yl-pyrimidine
88747
2-piperazin-1-ylpyrimidine
C1CN(CCN1)C2=NC=CC=N2
20980-22-7
C8H12N4
164.21
MFCD00040742
MRBFGEHILMYPTF-UHFFFAOYSA-N
2-1-piperazinyl pyrimidine,1-2-pyrimidyl piperazine,2-piperazin-1-yl pyrimidine,1-2-pyrimidinyl piperazine,2-piperazinopyrimidine,campiron,pyrimidine, 2-1-piperazinyl,campirone,kampirone,2-piperazin-1-yl-pyrimidine
88747
2-piperazin-1-ylpyrimidine
C1CN(CCN1)C2=NC=CC=N2
Boiling Point 96°C to 97°C (0.2mmHg)
Melting Point 29°C to 36°C
Density 1.0700g/mL
Flash Point >110°C
Specific Gravity 1.07
Refractive Index 1.5850 to 1.588
Quantity 5g
Color Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Beilstein 25, IV, 2083
Formula Weight 164.21
Physical Form Liquid After Melting
Percent Purity ≥98.0%
Chemical Name or Material 2-(1-Piperazinyl)pyrimidine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger