Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1-Piperidino)aniline, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass Bottle
39643-31-7
C11H16N2
176.263
MFCD00047467
OYECAJPUPWFCSL-UHFFFAOYSA-N
2-piperidinoaniline,2-piperidin-1-yl aniline,2-piperidin-1-yl-phenylamine,benzenamine, 2-1-piperidinyl,n-2-aminophenyl piperidine,2-1-piperidinyl aniline,2-1-piperidino aniline,1-2-aminophenyl piperidine,2-piperidylphenylamine,2-piperdinoaniline
458795
2-piperidin-1-ylaniline
C1CCN(CC1)C2=CC=CC=C2N
39643-31-7
C11H16N2
176.263
MFCD00047467
OYECAJPUPWFCSL-UHFFFAOYSA-N
2-piperidinoaniline,2-piperidin-1-yl aniline,2-piperidin-1-yl-phenylamine,benzenamine, 2-1-piperidinyl,n-2-aminophenyl piperidine,2-1-piperidinyl aniline,2-1-piperidino aniline,1-2-aminophenyl piperidine,2-piperidylphenylamine,2-piperdinoaniline
458795
2-piperidin-1-ylaniline
C1CCN(CC1)C2=CC=CC=C2N
Packaging Glass Bottle
Infrared Spectrum Authentic
Quantity 5g
Boiling Point 96.0°C (1.0 mmHg)
Color Brown
Melting Point 44.0°C to 48.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 176.26
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-(1-Piperidino)aniline, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning