Alfa Aesar™ 2-(1-Piperidinyl)-5-(trifluoromethyl)aniline

Manufacturer: Alfa Aesar™ H5072303

Quantity 1g

 View more versions of this product

Catalog No. AAH5072303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1496-40-8
    Molecular Formula C12H15F3N2
    Formula Weight 244.26
    MDL Number MFCD00042161
    UN Number UN2811
    Synonym 3-Amino-4-(1-piperidinyl)benzotrifluoride
    Chemical Name or Material 2-(1-Piperidinyl)-5-(trifluoromethyl)aniline

    Specifications

    Formula Weight 244.26
    Molecular Formula C12H15F3N2
    MDL Number MFCD00042161
    CAS 1496-40-8
    UN Number UN2811
    Synonym 3-Amino-4-(1-piperidinyl)benzotrifluoride
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H332-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Quantity 1g
    Chemical Name or Material 2-(1-Piperidinyl)-5-(trifluoromethyl)aniline

    GHS H Statement
    H301-H311-H332-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit