Alfa Aesar™ 2-(1-Piperidinyl)benzonitrile, 97%

Manufacturer: Alfa Aesar™ A1178406

Quantity 5g

 View more versions of this product

Catalog No. AAA1178406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 72752-52-4
    Molecular Formula C12H14N2
    Formula Weight 186.26
    MDL Number MFCD00049221
    UN Number UN3077
    Beilstein 1243909
    Synonym 1-(2-Cyanophenyl)piperidine; 2-(1-Piperidino)benzonitrile
    Chemical Name or Material 2- (1-Piperidinyl)benzonitrile

    Specifications

    Formula Weight 186.26
    MDL Number MFCD00049221
    CAS 72752-52-4
    Beilstein 1243909
    Synonym 1-(2-Cyanophenyl)piperidine; 2-(1-Piperidino)benzonitrile
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Boiling Point 122° to 125°C (1mmHg)
    Molecular Formula C12H14N2
    UN Number UN3077
    Melting Point 41° to 45°C
    Quantity 5g
    Chemical Name or Material 2- (1-Piperidinyl)benzonitrile
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit