Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[[(1E)-2-(Methoxycarbonyl)-1-methylvinyl]amino]-(2R)-2-cyclohexa-1,4-dienylacetic acid, sodium salt, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
85896-06-6
C14H19KNO4
304.407
MFCD00196811
SOILCJDBRKTYJP-BTQNPOSSSA-N
131886144
(2R)-2-cyclohexa-1,4-dien-1-yl-2-[(4-ethoxy-4-oxobut-2-en-2-yl)amino]acetic acid;potassium
CCOC(=O)C=C(C)NC(C1=CCC=CC1)C(=O)O.[K]
85896-06-6
C14H19KNO4
304.407
MFCD00196811
SOILCJDBRKTYJP-BTQNPOSSSA-N
131886144
(2R)-2-cyclohexa-1,4-dien-1-yl-2-[(4-ethoxy-4-oxobut-2-en-2-yl)amino]acetic acid;potassium
CCOC(=O)C=C(C)NC(C1=CCC=CC1)C(=O)O.[K]
Quantity 5g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H7CH(COONa)NHC(CH3)CHC(O)OCH3
Formula Weight 273.26
Chemical Name or Material 2-[[(1E)-2-(Methoxycarbonyl)-1-methylvinyl]amino]-(2R)-2-cyclohexa-1, 4-dienylacetic acid, sodium salt, Na salt, 98%

EINECSNumber : 288-783-7

TSCA : TSCA