Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(1H-Benzotriazol-1-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate, 97+%

Peptide coupling reagent, in solid phase synthesis

Manufacturer:  Alfa Aesar™ 43339-06

Quantity 5g

 View more versions of this product

Catalog No. AA4333906

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

125700-67-6
321.1
TBTU
C11H16N5O·BF4
MFCD00077413
2- (1H-Benzotriazol-1-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate

Specifications

205°C (dec.)
125700-67-6
C11H16N5O·BF4
MFCD00077413
2- (1H-Benzotriazol-1-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate
5g
Powder
321.1
TBTU
≥97%

Safety and Handling

GHS H Statement
H315-H319-H335

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit