Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2,2,3'-Tetrafluoroacetophenone, 98%

Click to view available options
Quantity:
1g
5g
708-64-5
C8H4F4O
192.113
MFCD00039236
NQSPTMFCJGKOQJ-UHFFFAOYSA-N
2,2,2,3'-tetrafluoroacetophenone,2,2,2-trifluoro-1-3-fluorophenyl ethanone,m-fluorotrifluoroacetophenone,acetophenone, 2,2,2,3'-tetrafluoro,2,2,2-trifluoro-1-3-fluorophenyl ethan-1-one,ethanone, 2,2,2-trifluoro-1-3-fluorophenyl,ethanone,2,2,2-trifluoro-1-3-fluorophenyl,acmc-20aol4,m,alpha,alpha,alpha-tetrafluoroacetophenone,2,2,2,3'-trtrafluoroacetophenone
69723
2,2,2-trifluoro-1-(3-fluorophenyl)ethanone
C1=CC(=CC(=C1)F)C(=O)C(F)(F)F
708-64-5
C8H4F4O
192.113
MFCD00039236
NQSPTMFCJGKOQJ-UHFFFAOYSA-N
2,2,2,3'-tetrafluoroacetophenone,2,2,2-trifluoro-1-3-fluorophenyl ethanone,m-fluorotrifluoroacetophenone,acetophenone, 2,2,2,3'-tetrafluoro,2,2,2-trifluoro-1-3-fluorophenyl ethan-1-one,ethanone, 2,2,2-trifluoro-1-3-fluorophenyl,ethanone,2,2,2-trifluoro-1-3-fluorophenyl,acmc-20aol4,m,alpha,alpha,alpha-tetrafluoroacetophenone,2,2,2,3'-trtrafluoroacetophenone
69723
2,2,2-trifluoro-1-(3-fluorophenyl)ethanone
C1=CC(=CC(=C1)F)C(=O)C(F)(F)F
Refractive Index 1.4440
Color Colorless to Yellow
Boiling Point 79°C (50mmHg)
Quantity 1g
Density 1.372g/mL
UN Number UN1224
Beilstein 1872788
Formula Weight 192.11
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2,2,2,3'-Tetrafluoroacetophenone
Show More Show Less

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.

TSCA : TSCA

storageNote1 : 2° to 8°C