Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-[2-(2-Chloroethoxy)ethoxy]ethanol 96.0+%, TCI America™
Click to view available options
Quantity:
25 g
100 g
500 g
Specifications
Color | Yellow |
Boiling Point | 232°C |
Quantity | 100 g |
Formula Weight | 168.62 |
Percent Purity | ≥96.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 2-[2-(2-Chloroethoxy)ethoxy]ethanol |
Safety and Handling
TSCA : No