Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Tribromoethanol, 99%, Thermo Scientific Chemicals

Catalog Number AAA1870606
Click to view available options
Quantity:
5 g
25 g
100 g
75-80-9
C2H3Br3O
282.757
MFCD00004671
YFDSDPIBEUFTMI-UHFFFAOYSA-N
tribromoethanol, avertin, bromethol, ethobrome, tribromethanol, ethobrom, basibrol, narcolan, narcotyl, narkolan
6400
2,2,2-tribromoethanol
C(C(Br)(Br)Br)O
75-80-9
C2H3Br3O
282.757
MFCD00004671
YFDSDPIBEUFTMI-UHFFFAOYSA-N
tribromoethanol, avertin, bromethol, ethobrome, tribromethanol, ethobrom, basibrol, narcolan, narcotyl, narkolan
6400
2,2,2-tribromoethanol
C(C(Br)(Br)Br)O

2,2,2-Tribromoethanol is utilized in organic synthesis of beta-amino alcohols. The pharmaceutical preparation of tribromoethanol serves as an anesthetic in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Tribromoethanol is utilized in organic synthesis of beta-amino alcohols. The pharmaceutical preparation of tribromoethanol serves as an anesthetic in medicine.

Solubility
Soluble in alcohol, ether, benzene and warmpetroleum ether. Partially soluble in water.

Notes
Light Sensitive. Store in cool and well ventilated area. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Melting Point 77°C to 80°C
Boiling Point 92°C to 93°C (10 mmHg)
Quantity 5 g
Beilstein 1733249
Sensitivity Light sensitive
Solubility Information Soluble in alcohol,ether,benzene and warmpetroleum ether. Partially soluble in water.
Formula Weight 282.76
Percent Purity 99%
Chemical Name or Material 2,2,2-Tribromoethanol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-903-1
RTECSNumber KM3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit