Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trichloroacetamide, 98+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
594-65-0
C2H2Cl3NO
162.394
MFCD00008009
UPQQXPKAYZYUKO-UHFFFAOYSA-N
trichloroacetamide,acetamide, 2,2,2-trichloro,amid kyseliny trichloroctove,acetamide, alpha-trichloro,unii-i8bl3305ro,amid kyseliny trichloroctove czech,2,2,2-trichloro-acetamide,acetamide, .alpha.-trichloro,.alpha.,.alpha.,.alpha.-trichloroacetamide,trichloro-acetamide
61144
2,2,2-trichloroacetamide
C(=O)(C(Cl)(Cl)Cl)N
594-65-0
C2H2Cl3NO
162.394
MFCD00008009
UPQQXPKAYZYUKO-UHFFFAOYSA-N
trichloroacetamide,acetamide, 2,2,2-trichloro,amid kyseliny trichloroctove,acetamide, alpha-trichloro,unii-i8bl3305ro,amid kyseliny trichloroctove czech,2,2,2-trichloro-acetamide,acetamide, .alpha.-trichloro,.alpha.,.alpha.,.alpha.-trichloroacetamide,trichloro-acetamide
61144
2,2,2-trichloroacetamide
C(=O)(C(Cl)(Cl)Cl)N
Packaging Glass bottle
Infrared Spectrum Authentic
Color White
Quantity 500g
Melting Point 139°C to 143°C
CAS Min % 98,98%
Assay Percent Range 98+%
Linear Formula CCl3CONH2
Beilstein 02, 211
Formula Weight 162.4
Physical Form Powder, Crystals and/or Chunks
Percent Purity ≥98%
Chemical Name or Material 2, 2, 2-Trichloroacetamide
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.