Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trichloroethanol, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
115-20-8
C2H3Cl3O
149.4
MFCD00004677
KPWDGTGXUYRARH-UHFFFAOYSA-N
trichloroethanol,trichlorethanol,trichloroethyl alcohol,ethanol, 2,2,2-trichloro,2,2,2-trichloro-1-ethanol,hydroxymethyl trichloromethane,2,2,2-trichloroethyl alcohol,2,2,2-trichloroethan-1-ol,unii-aw835aj62n,beta-trichloroethanol
8259
CHEBI:28094
2,2,2-trichloroethanol
C(C(Cl)(Cl)Cl)O
115-20-8
C2H3Cl3O
149.4
MFCD00004677
KPWDGTGXUYRARH-UHFFFAOYSA-N
trichloroethanol,trichlorethanol,trichloroethyl alcohol,ethanol, 2,2,2-trichloro,2,2,2-trichloro-1-ethanol,hydroxymethyl trichloromethane,2,2,2-trichloroethyl alcohol,2,2,2-trichloroethan-1-ol,unii-aw835aj62n,beta-trichloroethanol
8259
CHEBI:28094
2,2,2-trichloroethanol
C(C(Cl)(Cl)Cl)O
Flash Point 88°C
Density 1.5500g/mL
Color Colorless to Yellow
Boiling Point 151.0°C
Quantity 100g
Infrared Spectrum Authentic
Melting Point 17.8°C
Packaging Glass bottle
Specific Gravity 1.55
Refractive Index 1.4885 to 1.4905
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CCl3CH2OH
Beilstein 01, 338
Fieser 03,295; 04,521; 06,605
Merck Index 15, 9803
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,ether and carbon,tetrachloride
Formula Weight 149.4
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2, 2, 2-Trichloroethanol, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause drowsiness or dizziness.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Rinse mouth.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 204-071-0

RTECSNumber : KM3850000

TSCA : TSCA