Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroacetamide, 97%, Acros Organics

Click to view available options
Quantity:
25g
250g
2.5kg
354-38-1
C2H2F3NO
113.04
MFCD00008008
NRKYWOKHZRQRJR-UHFFFAOYSA-N
trifluoroacetamide,acetamide, 2,2,2-trifluoro,unii-36mu16byfk,n-trifluoroacetamide,2,2,2-trifluoro-acetamide,36mu16byfk,trifluroacetamide,trifluoro-acetamide,snylgqhjptadtaetp@,pubchem12622
67717
2,2,2-trifluoroacetamide
C(=O)(C(F)(F)F)N
354-38-1
C2H2F3NO
113.04
MFCD00008008
NRKYWOKHZRQRJR-UHFFFAOYSA-N
trifluoroacetamide,acetamide, 2,2,2-trifluoro,unii-36mu16byfk,n-trifluoroacetamide,2,2,2-trifluoro-acetamide,36mu16byfk,trifluroacetamide,trifluoro-acetamide,snylgqhjptadtaetp@,pubchem12622
67717
2,2,2-trifluoroacetamide
C(=O)(C(F)(F)F)N
Boiling Point 162°C to 164°C
Packaging Plastic Bottle
Melting Point 59°C to 70°C
Color White to Yellow or Beige
Quantity 2.5kg
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CF3CONH2
Beilstein 02, II, 186
Solubility Information Solubility in water: 460g/l (20°C). Other solubilities: soluble in alcohol,ether,partially soluble in chloroform
Formula Weight 113.04
Physical Form Crystalline Powder or Crystals
Percent Purity 97%
Chemical Name or Material 2, 2, 2-Trifluoroacetamide
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-559-9