Alfa Aesar™ 2,2,2-Trifluoroethanol-d2, 98% (Isotopic)

NMR Solvent

Manufacturer: Alfa Aesar™ 4236206

Quantity 5g

 View more versions of this product

Catalog No. AA4236206

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Chemical Identifiers

    CAS 132248-58-9
    Formula Weight 102.02
    MDL Number MFCD00082609
    UN Number UN1992
    Chemical Name or Material 2,2,2-Trifluoroethanol-d2, Isotopic

    Specifications

    Formula Weight 102.02
    Health Hazard 2 GHS H Statement
    H331-H318-H226-H302-H312-H315-H335
    Toxic if inhaled.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1992
    Sensitivity Hygroscopic
    Quantity 5g
    MDL Number MFCD00082609
    CAS 132248-58-9
    Boiling Point 77° to 80°C
    Melting Point −44°C
    Health Hazard 1 Danger
    Physical Form Liquid
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
    Chemical Name or Material 2,2,2-Trifluoroethanol-d2, Isotopic
    Assay 98%

    GHS H Statement
    H331-H318-H226-H302-H312-H315-H335
    Toxic if inhaled.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit