Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2,2-Trifluoroethanol-d3, 100%(Isotopic)

NMR Solvent

Manufacturer:  Alfa Aesar™ 4236403

Quantity 1g

 View more versions of this product

Catalog No. AA4236403


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

77253-67-9
MFCD00037682
2,2,2-Trifluoroethanol-d3, Isotopic
103.06
UN1992

Specifications

77253-67-9
1.45g/mL
−44°C
1g
MFCD00037682
2,2,2-Trifluoroethanol-d3, Isotopic
Hygroscopic
77° to 80°C
Liquid
103.06
UN1992
100%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H318-H226-H302-H312-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit